Learn More
Cayman Chemical (±)trans-2,5-bis-(3,4,5-Trimethoxyphenyl)-1,3-dioxolane, Purity: ≥98%, CAS: 116673-45-1, 10 mg
SDP
Catalog No. 502896483
Encompass_Preferred
trans-BTP Dioxolane is a PAF receptor antagonist. This compound demonstrates competitive antagonism of PAF in a rabbit washed platelet assay (Ki = 0.3 µM).{824}
Catalog No. 50-289-6483 Supplier Cayman Chemical Supplier No. 6095010 MG
trans-BTP Dioxolane is a PAF receptor antagonist. This compound demonstrates competitive antagonism of PAF in a rabbit washed platelet assay (Ki = 0.3 µM).{824}