Learn More
Cayman Chemical trans-2-Octenoyl-L-carnitine, Purity: ≥95%, CAS: 152064-94-3, 10 mg
SDP
Catalog No. 502990083
Encompass_Preferred
A synthetic carnitine; has been used as a standard for the quantification of carnitines in human urine
Catalog No. 50-299-0083 Supplier Cayman Chemical Supplier No. 3099510 MG
A synthetic carnitine; has been used as a standard for the quantification of carnitines in human urine