Learn More
Cayman Chemical TrnscrIptIon Factr Oct4 96 1ea
SDP
Catalog No. 502894204
Encompass_Preferred
A peptide MC1R antagonist (IC50 17 nM) selectively binds to MC1R over MC3R and MC5R (IC50s 3900 and 1000 nM respectively) as well as MC4R at 10 UM increases the latency to tail withdrawal in the tail-flick test in female but not male mice in a model of opioid-induced hyperalgesia at 7.5 UG/animal i.c.v. reduces protein levels of merlin in the cortex reduces the number of surviving neurons in the cortex and hippocampal CA2 region and increases the number of apoptotic cells in the cortex in rats 24 hours after injury in a model of controlled cortical impact-induced TBI at 1 nmol/animal i.c.v.
Catalog No. 50-289-4204 Supplier Cayman Chemical Supplier No. 6010841 EA
A peptide MC1R antagonist (IC50 17 nM) selectively binds to MC1R over MC3R and MC5R (IC50s 3900 and 1000 nM respectively) as well as MC4R at 10 UM increases the latency to tail withdrawal in the tail-flick test in female but not male mice in a model of opioid-induced hyperalgesia at 7.5 UG/animal i.c.v. reduces protein levels of merlin in the cortex reduces the number of surviving neurons in the cortex and hippocampal CA2 region and increases the number of apoptotic cells in the cortex in rats 24 hours after injury in a model of controlled cortical impact-induced TBI at 1 nmol/animal i.c.v.