Learn More
Cayman Chemical TrnscrIPTN FCTR Oct4 Pos 150uL
SDP
Catalog No. 502894203
Encompass_Preferred
An ERa degrader (IC50 4.1 nM) inhibits estradiol-induced ERa transcriptional activity in a reporter assay using T47D cells at 1 or 10 nM decreases the proliferation of MCF-7 and T47D cells and tamoxifen-resistant MCF-7 and T47D cells (IC50s 0.0032 0.0062 0.069 and 0.037 nM respectively)
Catalog No. 50-289-4203 Supplier Cayman Chemical Supplier No. 601081150 UL
An ERa degrader (IC50 4.1 nM) inhibits estradiol-induced ERa transcriptional activity in a reporter assay using T47D cells at 1 or 10 nM decreases the proliferation of MCF-7 and T47D cells and tamoxifen-resistant MCF-7 and T47D cells (IC50s 0.0032 0.0062 0.069 and 0.037 nM respectively)