Learn More
Cayman Chemical Transcrocetin, Purity: ≥98%, CAS: 27876-94-4, 50 mg
SDP
Catalog No. 502935876
Encompass_Preferred
A natural apocarotenoid with diverse biological activities; scavenges DPPH radicals (IC50 = 17.8 μg/ml); inhibits growth of MKN28 stomach, MCF-7 breast, and Caco-2 colon cancer cell lines (IC50s = 53, 63, and 103 μM, respectively); protects primary rat microglial cells from LPS-induced death and decreases LPS-induced production of intracellular ROS, TNF-α, IL-1β, and NF-κB (20 μM); increases GSH, CAT, CK, and LDH levels in cardiac tissue in a rat model of isoproterenol-induced myocardial infarction (100 mg/kg); increases the pressure threshold and reaction time to mechanical and thermal allodynia, respectively, in mice with spared nerve injury (30 mg/kg, intrathecal)
Catalog No. 50-293-5876 Supplier Cayman Chemical Supplier No. 2421750 MG
A natural apocarotenoid with diverse biological activities; scavenges DPPH radicals (IC50 = 17.8 μg/ml); inhibits growth of MKN28 stomach, MCF-7 breast, and Caco-2 colon cancer cell lines (IC50s = 53, 63, and 103 μM, respectively); protects primary rat microglial cells from LPS-induced death and decreases LPS-induced production of intracellular ROS, TNF-α, IL-1β, and NF-κB (20 μM); increases GSH, CAT, CK, and LDH levels in cardiac tissue in a rat model of isoproterenol-induced myocardial infarction (100 mg/kg); increases the pressure threshold and reaction time to mechanical and thermal allodynia, respectively, in mice with spared nerve injury (30 mg/kg, intrathecal)