Learn More
Cayman Chemical trans-β-Farnesene, Purity: ≥98%, CAS: 18794-84-8, 250 mg
SDP
Catalog No. 502990970
Encompass_Preferred
A volatile sesquiterpene hydrocarbon with pheromonal and kairomonal activities; produced by aphids in response to predator or parasite attack; induces dispersion of 50% of M. persicae aphids from a cluster at 0.2 ng; attracts larvae and adults of A. bipunctata at >2 μg
Catalog No. 50-299-0970 Supplier Cayman Chemical Supplier No. 31502250 MG
A volatile sesquiterpene hydrocarbon with pheromonal and kairomonal activities; produced by aphids in response to predator or parasite attack; induces dispersion of 50% of M. persicae aphids from a cluster at 0.2 ng; attracts larvae and adults of A. bipunctata at >2 μg