Learn More
Cole-Parmer Argos Technologies Transfer Pipette, 1.5 mL, Graduated to 0.3 mL; 500/Box
Catalog No. 501960565 Shop All Cole-Parmer Products
Encompass_Preferred
These pipettes are low-density polyethylene, which makes these disposable. The pipettes have built-in bulbs, and a perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops. Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use with a temperature range of -50 to 90C and can be gas sterilized.
Catalog No. 50-196-0565 Supplier Cole-Parmer Supplier No. 0622613
These pipettes are low-density polyethylene, which makes these disposable. The pipettes have built-in bulbs, and a perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops. Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use with a temperature range of -50 to 90C and can be gas sterilized.