Learn More
MTC Bio 5ml Large Bulb sterile, Individually Wrap
Supplier:  MTC Bio P411111
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass & exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) & are ideal for microbiology, blood banking, hematology, immunology, & urinalysis. Sterile pipettes are excellent for tissue culture application. Their low-affinity surface reduces sample, cell or protein loss. Whether you make your selection based on length, build draw, pipette capacity or number of drops per ml, one of these ten designs is likely to be suitable for your application. Some have ultra-fine extended tips (P4113, P4117,P4121, P4122, P4133) that allow for extremely precise dispensing & access into the narrowest vessels.
Catalog No. 50-112-7213