Learn More
MTC Bio Transfer Pipettes, 500/CS
Catalog No. 501871067
Encompass_Preferred
This item has a minimum qty of 4 per supplier requirements.
Transfer pipette, 7ml, large bulb, graduated to 3mL, sterile, peel packs of 20
Catalog No. 50-187-1067 Supplier MTC Bio Supplier No. P411414
This item has a minimum qty of 4 per supplier requirements.
Transfer pipette, 7ml, large bulb, graduated to 3mL, sterile, peel packs of 20