Learn More
Cayman Chemical TrnsfRrIn Receptr ProteIn 50uL
SDP
Catalog No. 502915515
Encompass_Preferred
A photosensitizer induces cell death in RIF-1 cells when used in combination with PDT at 0.06 and 1 UM induces IL-1 receptor internalization and NF-KB activation following light exposure in HCT116 cells
Catalog No. 50-291-5515 Supplier Cayman Chemical Supplier No. 3708050 UL
A photosensitizer induces cell death in RIF-1 cells when used in combination with PDT at 0.06 and 1 UM induces IL-1 receptor internalization and NF-KB activation following light exposure in HCT116 cells