Learn More
Cayman Chemical TrnsfRrIn RECPTR ProteIn 100ug
SDP
Catalog No. 502910032
Encompass_Preferred
Immunogen: Purified membrane fractions from OCI-LY7 cells • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) TfR1 • Applications: IF • MW: ~85 kDa
Catalog No. 50-291-0032 Supplier Cayman Chemical Supplier No. 36655100 UG
Immunogen: Purified membrane fractions from OCI-LY7 cells • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) TfR1 • Applications: IF • MW: ~85 kDa