Learn More
Cayman Chemical Transferrin Receptor Protein 1/CD71 Monoclonal Antibody (PE) (Clone 04), 25 test
SDP
Catalog No. 502915022
Encompass_Preferred
Immunogen: Recombinant human TfR1 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) TfR1 • Applications: FC
Catalog No. 50-291-5022 Supplier Cayman Chemical Supplier No. 3708125 TEST
Immunogen: Recombinant human TfR1 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) TfR1 • Applications: FC