Learn More
Cayman Chemical TrnsglutamInase 2 RabbIt Mon
SDP
Catalog No. 502915108
Encompass_Preferred
An inhibitor of ALK5 (IC50 2.5 nM) selective for ALK5 over p38 MAPK (IC50 12 UM) reduces renal fibrosis in a model of unilateral ureteral obstruction using transgenic Col1a1-Luc Tg rats at 1 3 and 10 mg/kg reduces proteinuria and glomerulosclerosis and improves renal function in heminephrectomized rats in a model of anti-Thy-1 glomerulonephritis at 0.3 and 1 mg/kg
Catalog No. 50-291-5108 Supplier Cayman Chemical Supplier No. 37090100 UL
An inhibitor of ALK5 (IC50 2.5 nM) selective for ALK5 over p38 MAPK (IC50 12 UM) reduces renal fibrosis in a model of unilateral ureteral obstruction using transgenic Col1a1-Luc Tg rats at 1 3 and 10 mg/kg reduces proteinuria and glomerulosclerosis and improves renal function in heminephrectomized rats in a model of anti-Thy-1 glomerulonephritis at 0.3 and 1 mg/kg