Learn More
Cayman Chemical TrnsglutamInase 2humnre 100ug
SDP
Catalog No. 502964453
Encompass_Preferred
Source: Active N-terminally His-tagged human TG2 protein (full length) purified from E. coli • MW: 79.4 kDa
Catalog No. 50-296-4453 Supplier Cayman Chemical Supplier No. 23595100 UG
Source: Active N-terminally His-tagged human TG2 protein (full length) purified from E. coli • MW: 79.4 kDa