Learn More
Cayman Chemical Transglutaminase Assay Stop Solution, 10 mL
SDP
Catalog No. 502942384
Encompass_Preferred
Transglutaminase Assay Stop Solution has been tested and formulated to work exclusively with Cayman’s Transglutaminase Activity Assay Kit (Item No. 702250).
Catalog No. 50-294-2384 Supplier Cayman Chemical Supplier No. 40054210 ML
Transglutaminase Assay Stop Solution has been tested and formulated to work exclusively with Cayman’s Transglutaminase Activity Assay Kit (Item No. 702250).