Learn More
Cayman Chemical Transglutaminase Assay Substrate 1, 1 ea
SDP
Catalog No. 502944014
Encompass_Preferred
Transglutaminase Assay Substrate 1 has been tested and formulated to work exclusively with Cayman’s Transglutaminase Activity Assay Kit (Item No. 702250).
Catalog No. 50-294-4014 Supplier Cayman Chemical Supplier No. 4005401 EA
Transglutaminase Assay Substrate 1 has been tested and formulated to work exclusively with Cayman’s Transglutaminase Activity Assay Kit (Item No. 702250).