Learn More
Electron Microscopy Sciences Transport Tubes, 500/PK
SDP
Supplier:  Electron Microscopy Sciences 6303228
Encompass_Preferred
These transparent polypropylene tubes are designed for the safe transport and storage of biological samples and materials. They are shatter-proof and can be exposed to freezing temperatures. The transport tubes are self-standing with a conical bottom. With a leak-resistant screw caps that can be sealed or removed with a single turn. Sterile Cap: Attached . Size: 10mL .
Catalog No. 50-209-0793