Learn More
Cayman Chemical trans-Vaccenic Acid methyl ester, Purity: ≥98%, CAS: 6198-58-9, 250 mg
SDP
Catalog No. 502935055
Encompass_Preferred
An esterified form of trans-vaccenic acid; has been used as a standard for the identification and quantification of fatty acids in Uruguayan Colonia cheese by GC-MS
Catalog No. 50-293-5055 Supplier Cayman Chemical Supplier No. 26731250 MG
An esterified form of trans-vaccenic acid; has been used as a standard for the identification and quantification of fatty acids in Uruguayan Colonia cheese by GC-MS