Learn More
Cayman Chemical trnsVaccenIc ACD methyl S 50mg
SDP
Catalog No. 502936287
Encompass_Preferred
An esterified form of trans-vaccenic acid; has been used as a standard for the identification and quantification of fatty acids in Uruguayan Colonia cheese by GC-MS
Catalog No. 50-293-6287 Supplier Cayman Chemical Supplier No. 2673150 MG
An esterified form of trans-vaccenic acid; has been used as a standard for the identification and quantification of fatty acids in Uruguayan Colonia cheese by GC-MS