Learn More
Cayman Chemical Tranexamic Acid-13C2,15N, Purity: ≥95%, CAS: 1292837-95-6, 1 mg
SDP
Catalog No. 502939423
Encompass_Preferred
An internal standard for the quantification of tranexamic acid by GC- or LC-MS
Catalog No. 50-293-9423 Supplier Cayman Chemical Supplier No. 288041 MG
An internal standard for the quantification of tranexamic acid by GC- or LC-MS