Learn More
Apexbio Technology LLC TRO 19622 22033-87-0 5mg
SDP
Catalog No. 502240197
Encompass_Preferred
TRO 19622 (CAS 22033-87-0) also known as Olesoxime is a cholesterol-like small molecule that modulates mitochondrial function contributing to cellular resilience under stress conditions Preclinical studies demonstrate that TRO 19622 attenuates neurodegeneration supports nerve function and facilitates neuroregeneration following traumatic nerve injury in both in vitro and in vivo models This compound is utilized in research exploring neuroprotection and mechanisms underlying neuronal survival with potential applications in models of neurodegenerative diseases and peripheral nerve trauma
This item is not returnable. View return policy
Catalog No. 50-224-0197 Supplier Apexbio Technology LLC Supplier No. B35835
TRO 19622 (CAS 22033-87-0) also known as Olesoxime is a cholesterol-like small molecule that modulates mitochondrial function contributing to cellular resilience under stress conditions Preclinical studies demonstrate that TRO 19622 attenuates neurodegeneration supports nerve function and facilitates neuroregeneration following traumatic nerve injury in both in vitro and in vivo models This compound is utilized in research exploring neuroprotection and mechanisms underlying neuronal survival with potential applications in models of neurodegenerative diseases and peripheral nerve trauma