Learn More
Med Vet International Trocar-Trocar I. M. Pin, 5/64" x 7", J0250B
Catalog No. 5000582550
Encompass_Preferred
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.
Catalog No. 50-005-82550 Supplier Med Vet International Supplier No. JOR250B
Highest quality type 316L stainless steel IM pins. Available in the points and diameters listed below. When ordering, please match diameter with point style number. Made in U.S.A.