Learn More
Braintree Scientific Trocars, Semi Blunt 45 degree beveled tip, 12g x 6""""
SDP
Catalog No. 501956069
Encompass
Trocars - Skin tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes subcutaneously. Our trocars are manufactured from thick walled medical grade stainless steel and have a semi-blunt 45 degree beveled tip. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6069 Supplier Braintree Scientific Supplier No. TRO126
Trocars - Skin tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes subcutaneously. Our trocars are manufactured from thick walled medical grade stainless steel and have a semi-blunt 45 degree beveled tip. **Sold for research purposes only and not for human use**