Learn More
Braintree Scientific Trocars,Tapered/Semi-Blunt Tip, 7g x 12"""" w/Stylet
SDP
Catalog No. 501956077
Encompass
Trocars - Skin tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes subcutaneously. Our trocars are manufactured from thick walled medical grade stainless steel and have a semi-blunt 45 degree beveled tip. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6077 Supplier Braintree Scientific Supplier No. TROSTY7B12
Trocars - Skin tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes subcutaneously. Our trocars are manufactured from thick walled medical grade stainless steel and have a semi-blunt 45 degree beveled tip. **Sold for research purposes only and not for human use**