Learn More
Med Vet International Trochlear Rasp 6.5", J0210, Each
Catalog No. 5000585187
Encompass_Preferred
Designed especially for remodeling the femoral trochlear groove in patella luxation. The varying cone- shape diameter allows the surgeon to make the patella fit precisely. Stainless steel. 17cm (6.5").
Catalog No. 50-005-85187 Supplier Med Vet International Supplier No. JOR210
Designed especially for remodeling the femoral trochlear groove in patella luxation. The varying cone- shape diameter allows the surgeon to make the patella fit precisely. Stainless steel. 17cm (6.5").