Learn More
Medchemexpress LLC Trodusquemine lactate | 1309370-86-2 | 98.8% | 1 ML
SDP
Catalog No. 5000231148
Encompass_Preferred
MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of 1 μM and approximately 200-fold selectivity over TCPTP. Supplied as a 10 mM stock solution (1 mL), it is intended for research use in biochemical and cell-based assays targeting PTP1B-regulated pathways.
  • Selective PTP1B inhibition, IC50 = 1 μM.
  • Approximately 200-fold selectivity over TCPTP.
  • Supplied as a 10 mM stock solution in 1 mL vial.
  • High purity as verified by HPLC.
  • CAS number 1309370-86-2 for substance identification.
  • Suitable for biochemical and cell-based assays.
Catalog No. 50-002-31148 Supplier Medchemexpress LLC Supplier No. HY12219A10MM 1ML
MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B), with an IC50 of 1 μM and approximately 200-fold selectivity over TCPTP. Supplied as a 10 mM stock solution (1 mL), it is intended for research use in biochemical and cell-based assays targeting PTP1B-regulated pathways.
  • Selective PTP1B inhibition, IC50 = 1 μM.
  • Approximately 200-fold selectivity over TCPTP.
  • Supplied as a 10 mM stock solution in 1 mL vial.
  • High purity as verified by HPLC.
  • CAS number 1309370-86-2 for substance identification.
  • Suitable for biochemical and cell-based assays.