Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trodusquemine lactate | 1309370-86-2 | 98.8% | 10 MG
SDP

Catalog No. 5000230724
Encompass_Preferred

MSI-1436 lactate (trodusquemine lactate) is a research-grade lactate salt that selectively inhibits protein-tyrosine phosphatase 1B (PTP1B). It is a non-competitive inhibitor with an in vitro IC50 of 1 μM and shows approximately 200-fold selectivity over TCPTP. The compound is supplied as a solid salt intended for metabolic and signaling research.

  • Selective, non-competitive PTP1B inhibitor (IC50 = 1 μM).
  • Approximately 200-fold selectivity over TCPTP.
  • Lactate salt supplied as a solid, suitable for in vitro and in vivo research.
  • Molecular weight 1000.17 g/mol; formula C40H78N4O8S.
  • Available in small research pack sizes, such as 10 mg.

Catalog No. 50-002-30724 Supplier Medchemexpress LLC Supplier No. HY12219A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MSI-1436 lactate (trodusquemine lactate) is a research-grade lactate salt that selectively inhibits protein-tyrosine phosphatase 1B (PTP1B). It is a non-competitive inhibitor with an in vitro IC50 of 1 μM and shows approximately 200-fold selectivity over TCPTP. The compound is supplied as a solid salt intended for metabolic and signaling research.

  • Selective, non-competitive PTP1B inhibitor (IC50 = 1 μM).
  • Approximately 200-fold selectivity over TCPTP.
  • Lactate salt supplied as a solid, suitable for in vitro and in vivo research.
  • Molecular weight 1000.17 g/mol; formula C40H78N4O8S.
  • Available in small research pack sizes, such as 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.