Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trodusquemine lactate | 1309370-86-2 | 98.8% | 25 MG
SDP

Catalog No. 5000231332
Encompass_Preferred

MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with reported in vitro potency of approximately 1 μM and about 200-fold selectivity over TCPTP (IC50 224 μM). The compound is supplied as the lactate salt and is used in biochemical and in vivo research targeting PTP1B-related signaling and metabolic regulation.

  • Selective PTP1B inhibitor with IC50 ≈ 1 μM
  • Approximately 200-fold selectivity over TCPTP (IC50 224 μM)
  • High reported purity (98.8%)
  • Molecular formula C40H78N4O8S; molecular weight ~1000.17 g/mol
  • Available in small milligram scales, including 25 MG
  • Suitable for biochemical and in vivo research applications

Catalog No. 50-002-31332 Supplier Medchemexpress LLC Supplier No. HY12219A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MSI-1436 lactate (trodusquemine lactate) is a selective, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with reported in vitro potency of approximately 1 μM and about 200-fold selectivity over TCPTP (IC50 224 μM). The compound is supplied as the lactate salt and is used in biochemical and in vivo research targeting PTP1B-related signaling and metabolic regulation.

  • Selective PTP1B inhibitor with IC50 ≈ 1 μM
  • Approximately 200-fold selectivity over TCPTP (IC50 224 μM)
  • High reported purity (98.8%)
  • Molecular formula C40H78N4O8S; molecular weight ~1000.17 g/mol
  • Available in small milligram scales, including 25 MG
  • Suitable for biochemical and in vivo research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.