Learn More
Troemner™ Bench Clamps
Catalog No. 02216297 Shop All Troemner Products
Change view
Click to view available options
Type:
Bench Clamp
Catalog No. Type
02-216-297 Bench Clamp
1 options
Catalog No. 02-216-297 Supplier Troemner™ Supplier No. 30392311
Add to Cart
Add to Cart
Bench clamp features an aluminum bracket with arm that fastens quickly and firmly to any convenient shelf.
Accepts 0.51 in. (13mm) rods that ordinary rings and clamps can attach to, leaving the bench surface clear for apparatus. Can accept rods vertically and horizontally.
TRUSTED_SUSTAINABILITY
Specifications
Type Bench Clamp
Description Accepts 0.51 in. (13 mm) rods