Learn More
Troemner™ Class 7 Aluminum Weights
Catalog No. 02301117 Shop All Troemner Products
Change view
Click to view available options
Weight (Metric):
2 mg
20 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
02-301-117 200 mg
02-301-115 2 mg
02-301-116 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-301-117 Supplier Troemner™ Supplier No. 30391011
Add to Cart
Add to Cart
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
Weight is not supplied in case. Protective case is available at an additional cost.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Weight
Classification Class 7
Material Aluminum
Weight (Metric) 200 mg