Learn More
Troemner™ Individual Analytical Precision Weights, UltraClass with NVLAP Certificate
Shop All Troemner ProductsDescription
Made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage.
- Exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to -195.5°C (-320°F)
- Fractional weights 500mg to 50mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are aluminum
- The UltraClass weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2
- This class is used as a reference standard for calibrating other reference standards and weights and is appropriate for calibrating high-precision analytical balances with a readability as low as 0.01mg
- Weights are supplied with a NIST/NVLAP Weight Calibration Certificate
- Weights supplied in a rugged, precision-fit, high quality polycarbonate case
Specifications
Specifications
| Product Type | One Piece Metric Analytical Sheet Weight |
| Certifications/Compliance | NVLAP, ISO9001 |
| Classification | UltraClass |
| Material | Aluminum |
| Weight (Metric) | 3 mg |
| Density | 2.7g/cm3 at 20°C |
| Mass | 3 mg |
| Shape | Flat with one end turned up for easy handling |
| Includes | Polycarbonate case |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.