Learn More
Sigma Aldrich Fine Chemicals Biosciences Troemner Lab Frame fume ho
Catalog No. 502909035
Encompass
Frame 36 x 50 in. (914 x 1270 mm)Base 12 in. (305 mm) wideComponents2 in. (51 mm) rods (qty 8)12 in. (305 mm) rods (qty 2)18 in. (457 mm) rods (qty 2)36 in. (914 mm) rods (qty 5)50 in. (1270 mm) rods (qty 4)Frame connectors (qty 34)Rod end connectors (qty 4)Lab-frame feet (qty 4)
Catalog No. 50-290-9035 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6943041KT
Frame 36 x 50 in. (914 x 1270 mm)Base 12 in. (305 mm) wideComponents2 in. (51 mm) rods (qty 8)12 in. (305 mm) rods (qty 2)18 in. (457 mm) rods (qty 2)36 in. (914 mm) rods (qty 5)50 in. (1270 mm) rods (qty 4)Frame connectors (qty 34)Rod end connectors (qty 4)Lab-frame feet (qty 4)