Learn More
Sigma Aldrich Fine Chemicals Biosciences Troemner Lab frame standar
Catalog No. 502905091
Encompass
Standard Lab-frame kits are available in five sizes and three lattice rod choicesTraditional center-less ground aluminumNon-corroding fiberglassTough stainless steelA range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
Catalog No. 50-290-5091 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6941421KT
Standard Lab-frame kits are available in five sizes and three lattice rod choicesTraditional center-less ground aluminumNon-corroding fiberglassTough stainless steelA range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).