Learn More
Troemner™ Support Stand Stabilizer Knobs
Catalog No. 02216353 Shop All Troemner Products
Change view
Click to view available options
Quantity:
2 Pk.
Catalog No. Quantity
02-216-353 2 Pk.
1 options
Catalog No. 02-216-353 Supplier Troemner™ Supplier No. 30400044
Add to Cart
Add to Cart
For securing the stand to a bench top
Support Stand Stabilizer Knobs can be added to the two additional threaded holes in the base legs to secure the stand to a bench top.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stabilizer Knobs
For Use With (Application) Secures stand to bench top
For Use With (Equipment) Support Stands
Quantity 2 Pk.