Learn More
Troemner™ Talboys™ Labjaws™ Chain Assembly
Catalog No. 02216368 Shop All Troemner Products
Change view
Click to view available options
Type:
Chain Assembly
Catalog No. Type
02-216-368 Chain Assembly
1 options
Catalog No. 02-216-368 Supplier Troemner™ Supplier No. 30531156
Add to Cart
Add to Cart
Replacement chain assembly for use on 717 Wall Bracket and 716 Bench Clamp.
Sturdy chain for extra security. Chain measures 49 in. (124.5cm).
TRUSTED_SUSTAINABILITY
Specifications
Type Chain Assembly