Learn More
Troemner™ Talboys™ Labjaws™ Support Stand for Double Buret Clamp
Catalog No. 02216298 Shop All Troemner Products
Change view
Click to view available options
For Use With (Equipment):
Labjaws
Catalog No. For Use With (Equipment)
02-216-298 Labjaws
1 options
Catalog No. 02-216-298 Supplier Troemner™ Supplier No. 30392312
Add to Cart
Add to Cart
Support stand with standard rod for use with the nickel-plated Double Buret Clamp (sold separately). Clamp unit attaches to the support rod with built-in reinforced hook connector. Support rod attaches to the porcelain base. Rod (Dia x L): 1.3 x 57.8cm, Stand (L x W x H): 17.8 x 33 x 2.5cm
  • For use with the nickel-plated Double Buret Clamp (sold separately), which holds any size buret from micro to 100mL capacity
TRUSTED_SUSTAINABILITY
Specifications
Material Nickel, Zinc, Aluminum, Porcelain
Includes Base and rod
Product Type Support Stand
For Use With (Equipment) Labjaws