Learn More
Troemner™ Talboys™ Labjaws™ Double Buret Clamp
Catalog No. 02300213 Shop All Troemner Products
Change view
Click to view available options
For Use With (Equipment):
Labjaws
Catalog No. For Use With (Equipment)
02-300-213 Labjaws
1 options
Catalog No. 02-300-213 Supplier Troemner™ Supplier No. 30392234
Add to Cart
Add to Cart
Nickel-plated zinc Double Buret Clamp holds any size buret from micro to 100mL capacity. Simply compress the scissor-like mechanism, insert buret and gently release to grip. Clamp only; rod and support stand available separately.
  • Numbers and graduation on buret remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release
  • Double Buret Clamp unit attaches to optional standard support rod with built-reinforced hook connector
  • Adjusting nut faces forward for easy use
  • Stainless steel or aluminum support rod attaches to optional porcelain base
TRUSTED_SUSTAINABILITY
Specifications
Type Double Burette Clamp
Material Nickel, Zinc
For Use With (Equipment) Labjaws