Learn More
Troemner™ Talboys™ Labjaws™ Holders for Clamps
Catalog No. 02217130 Shop All Troemner Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
02-217-130 Stainless Steel
1 options
Catalog No. 02-217-130 Supplier Troemner™ Supplier No. 30392228
Add to Cart
Add to Cart
Stainless steel, two-rod holder with center swivel capacity allows tilting of clamps at any angle in parallel planes.
Outside adjustments screw allows close proximity between items being held. Stainless steel electro-polished finish.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Includes Outside adjustment screw
Product Type Swivel Clamp Holder
Quantity Each
Weight (English) 1 lb.
Weight (Metric) 0.45 kg