Learn More
Troemner™ Talboys™ Labjaws™ Support Clamps
Catalog No. 02216352 Shop All Troemner Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
02-216-352 Stainless Steel
1 options
Catalog No. 02-216-352 Supplier Troemner™ Supplier No. 30400043
Add to Cart
Add to Cart
Standard support holder constructed of strong stainless steel.
  • Equipped with over sized, easy to grip knobs for precise tightening
  • Accommodates 1/2 in. (12.7mm) diameter shaft of a motor mounting rod and 5/8 in. (16mm) diameter support stand rod
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Diameter (English) Rod 0.625 in.
Diameter (Metric) Rod 1.59 cm
Product Type Support Holder
Quantity Each