Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Universal Stirrer Mounting Brackets

Catalog No. 02216263 Shop All Troemner Products
Change view
Click to view available options
Capacity (Metric):
2.27 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-216-263 2.27 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-216-263 Supplier Troemner™ Supplier No. 30392265
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use to mount stirrers and other devices to lab-frames.

Aluminum mounting bracket with attachment screws.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 5 lb.
Capacity (Metric) 2.27 kg
For Use With (Application) Use to mount stirrers and other devices to lab-frames
Includes Attachment screws
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.