Learn More
Cayman Chemical Troglitazone, Purity: ≥98%, CAS: 97322-87-7, 50 mg
SDP
Catalog No. 502894815
Encompass_Preferred
A selective agonist of PPARγ (EC50s = 0.78 and 0.55 μM for the mouse and human receptors, respectively, in a transactivation assay); selective for PPARγ over PPARα and PPARδ, at which it is inactive at ≤10 μM; exhibits antihyperglycemic and antihyperlipidemic properties in Zucker diabetic fatty rats, reducing plasma glucose and triglyceride levels by 61 and 87% respectively, at 500 mg/kg twice per day; induces cell cycle arrest at the G1 phase in SK-HEP-1 and Hep3B hepatocellular carcinoma cells at 10 μM and increases apoptosis in these cells at ≥30 μM
Catalog No. 50-289-4815 Supplier Cayman Chemical Supplier No. 7175050 MG
A selective agonist of PPARγ (EC50s = 0.78 and 0.55 μM for the mouse and human receptors, respectively, in a transactivation assay); selective for PPARγ over PPARα and PPARδ, at which it is inactive at ≤10 μM; exhibits antihyperglycemic and antihyperlipidemic properties in Zucker diabetic fatty rats, reducing plasma glucose and triglyceride levels by 61 and 87% respectively, at 500 mg/kg twice per day; induces cell cycle arrest at the G1 phase in SK-HEP-1 and Hep3B hepatocellular carcinoma cells at 10 μM and increases apoptosis in these cells at ≥30 μM