Learn More
AdipoGen Troglitazone (5 mg)
SDP
Supplier:  AdipoGen AGCR13565M005
Encompass_Preferred
Chemical. CAS: 97322-87-7. Formula: C24H27NO5S. MW: 441.5. Potent and selective PPARgamma agonist with no activation of PPARalpha and PPARdelta. Anti-diabetic agent with potent glucose-lowering effects and decreasing insulin resistance in adipose tissues. Stimulates adipogenesis. Apoptosis and cell cycle arrest inducer in several cancer cell lines. Anti-inflammatory. Inhibits vascular smooth muscle cell (VSMC) migration.
Catalog No. 50-205-3932