Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trolamine, NF, 99-107.4%, Spectrum™ Chemical
SDP

Catalog No. 18607603 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-603 500 mL Amber Glass Bottle
18-607-604 6 x 500 mL Amber Glass Bottle
18-607-605 4 L Amber Glass Bottle
18-607-606 4 x 4 L Amber Glass Bottle
18-607-607 20 L Amber Glass Bottle
18-607-608 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-603 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TR143500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

TR143, 102-71-6, C6H15NO3

Spectrum™ Chemical Trolamine, NF is used as an emulsifying excipient in hand and body lotions, shaving creams, soaps, and shampoos. All Spectrum Chemical NF products, including all NF Trolamine products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

CAS Max % 1
Boiling Point 335°C / 635°F
Ignition Residue 0.0005
Packaging Amber Glass Bottle
Refractive Index 1.481-1.486
Quantity 500 mL
Percent Purity 99 to 107.4%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Trolamine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.