Learn More
Medchemexpress LLC HY-108881 1mg , Troleandomycin CAS:2751-09-9 Purity:>98%
SDP
Catalog No. 502261648
Encompass_Preferred
HY-108881 1mg Troleandomycin CAS: 2751-09-9 Troleandomycin (Triacetyloleandomycin), a macrolide acrolide antibiotic, is a selective CYP3A inhibitor. Troleandomycin is an oral corticosteroid for asthma study. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1648 Supplier Medchemexpress LLC Supplier No. HY1088811MG
HY-108881 1mg Troleandomycin CAS: 2751-09-9 Troleandomycin (Triacetyloleandomycin), a macrolide acrolide antibiotic, is a selective CYP3A inhibitor. Troleandomycin is an oral corticosteroid for asthma study. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.