Learn More
Medchemexpress LLC HY-101445 500mg Medchemexpress, Trolox CAS:53188-07-1 Purity:98%
SDP
Catalog No. 501636960
Encompass_Preferred
Medchemexpress,HY-101445 500mg Trolox CAS:53188-07-1 Formula:C14H18O4 Troloxa is a cell-permeable and water-soluble vitamin E analogue and is a powerful antioxidant. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6960 Supplier Medchemexpress LLC Supplier No. HY101445500MG
Medchemexpress,HY-101445 500mg Trolox CAS:53188-07-1 Formula:C14H18O4 Troloxa is a cell-permeable and water-soluble vitamin E analogue and is a powerful antioxidant. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.