Learn More
Enzo Life Sciences Trolox (100mg). CAS: 53188-07-1; 56305-04-5
SDP
Supplier:  Enzo Life Sciences ALX270267M100
Encompass_Preferred
Cell permeable derivative of vitamin E. Antioxidant. Inhibits peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Prevents lipid peroxidation, cytotoxicity and apoptosis induced by cytochrome P450 2E1 in HEP G2 cells. Alternative name: 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid. Formula: C14H18O4. MW: 250.3. Purity: ≥96.5% (Titration (T) NaOH 0.1M). Solubility: Soluble in 100% ethanol (25mg/ml) or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-8453