Learn More
Medchemexpress LLC HY-101445 1g , Trolox CAS:53188-07-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014451G
Encompass_Preferred
HY-101445 1g Trolox CAS: 53188-07-1 Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0400