Learn More
AdipoGen Trolox,Chemical. CAS: 53188-07-1
SDP
Catalog No. 501176132
Encompass_Preferred
Trolox, AdipoGen Life Sciences. Chemical. CAS: 53188-07-1. Formula: C14H18O4. MW: 250.3. Cell permeable and water soluble derivative of vitamin E. Potent antioxidant. Efficiently prevents formation of different reactive oxygen species, such as singlet oxygen (1O2), superoxide anion (O2-) or hydrogen peroxide (H2O2). Commonly used as a standard or positive control in different antioxidant assays, such as ABTS decolorization and FRAP assays, measuring Trolox equivalent antioxidant capacity (TEAC). TEAC assays are used to measure antioxidant capacity of foods, beverages and supplements. Used in biological or biochemical applications to reduce oxidative stress or damage, such as inhibiting peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Cytoprotective with low cytotoxicity for different cell lines. Shown to have anticancer activity and to be effective as adjunctive agent in cancer treatments. 53188-07-1
Catalog No. 50-117-6132 Supplier AdipoGen Supplier No. AGCR13639G001
Trolox, AdipoGen Life Sciences. Chemical. CAS: 53188-07-1. Formula: C14H18O4. MW: 250.3. Cell permeable and water soluble derivative of vitamin E. Potent antioxidant. Efficiently prevents formation of different reactive oxygen species, such as singlet oxygen (1O2), superoxide anion (O2-) or hydrogen peroxide (H2O2). Commonly used as a standard or positive control in different antioxidant assays, such as ABTS decolorization and FRAP assays, measuring Trolox equivalent antioxidant capacity (TEAC). TEAC assays are used to measure antioxidant capacity of foods, beverages and supplements. Used in biological or biochemical applications to reduce oxidative stress or damage, such as inhibiting peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Cytoprotective with low cytotoxicity for different cell lines. Shown to have anticancer activity and to be effective as adjunctive agent in cancer treatments. 53188-07-1