Learn More
Medchemexpress LLC HY-101445 5g Medchemexpress, Trolox CAS:53188-07-1 Purity:>98%
SDP
Catalog No. 501932133
Encompass_Preferred
Medchemexpress, HY-101445 5g Trolox CAS:53188-07-1 Trolox is a vitamin E analogue and is a powerful antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2133 Supplier Medchemexpress LLC Supplier No. HY1014455G
Medchemexpress, HY-101445 5g Trolox CAS:53188-07-1 Trolox is a vitamin E analogue and is a powerful antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.