Learn More
Abcam Trolo x , Antio x idant vitamin E derivative, 500MG
Catalog No. 502707310
Encompass_Preferred
MW 250.29 g/mol, Purity >98%. Antioxidant vitamin E derivative. Exhibits lipid peroxidizing, DPPH° scavenging and HO° scavenging activity. Regularly used as an antioxidant standard. Cell-permeable and water soluble.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7310 Supplier Abcam Supplier No. AB120747500MG
MW 250.29 g/mol, Purity >98%. Antioxidant vitamin E derivative. Exhibits lipid peroxidizing, DPPH° scavenging and HO° scavenging activity. Regularly used as an antioxidant standard. Cell-permeable and water soluble.
The product is subject to the following: Abcam Restricted Use Statement